Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Na+, K+ ATPase β3 Mouse anti-Human, Unlabeled, Clone: 46, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610992

 View more versions of this product

Catalog No. BDB610992


Add to cart

Description

Description

Na+,K+-ATPase is an integral plasma membrane protein that maintains the intracellular concentrations of Na+ and K+ ions. It is a P class pump that is composed of two α and two β transmembrane subunits. It is also thought to contain a γ subunit. The 120kDa α subunit represents the catalytic site. Although the 50kDa β subunit mediates proper folding of newly synthesized α subunits, it is not directly involved in ion
transfer. There are four known α isoforms (α1, α2, α3, and α4) and three β isoforms (β1, β2, and β3). The Na+,K+-ATPase is ubiquitously expressed in the plasma membrane. However, the individual subunits have different tissue distributions: α1 is ubiquitous; α2 is in skeletal muscle, brain, and heart; α3 is in brain and heart; α4 is present in testis and skeletal muscle; β1 is in most tissues; and β2 is primarily in neural tissues. Different combinations of the α and β subunits can alter the kinetics of enzyme activity. The Na+,K+-ATPase transfers three Na+ ions out of, and two K+ ions into, the cell upon hydrolysis of one ATP molecule. Maintenance of intracellular Na+,K+ levels is necessary for volume regulation, action potentials, and secondary active transport.

Immunofluorescence, Western Blotting

Specifications

Specifications

Na+, K+ ATPase β3
46
Unconjugated
Affinity Purified
Human Na+
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.