Learn More
Na+, K+ ATPase β3 Mouse anti-Human, Unlabeled, Clone: 46, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610993
Description
Na+,K+-ATPase is an integral plasma membrane protein that maintains the intracellular concentrations of Na+ and K+ ions. It is a P class pump that is composed of two α and two β transmembrane subunits. It is also thought to contain a γ subunit. The 120kDa α subunit represents the catalytic site. Although the 50kDa β subunit mediates proper folding of newly synthesized α subunits, it is not directly involved in ion
transfer. There are four known α isoforms (α1, α2, α3, and α4) and three β isoforms (β1, β2, and β3). The Na+,K+-ATPase is ubiquitously expressed in the plasma membrane. However, the individual subunits have different tissue distributions: α1 is ubiquitous; α2 is in skeletal muscle, brain, and heart; α3 is in brain and heart; α4 is present in testis and skeletal muscle; β1 is in most tissues; and β2 is primarily in neural tissues. Different combinations of the α and β subunits can alter the kinetics of enzyme activity. The Na+,K+-ATPase transfers three Na+ ions out of, and two K+ ions into, the cell upon hydrolysis of one ATP molecule. Maintenance of intracellular Na+,K+ levels is necessary for volume regulation, action potentials, and secondary active transport.
Immunofluorescence, Western Blotting
Specifications
Na+, K+ ATPase β3 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human Na+, K+ ATPase β3 aa. 124-243 | |
150 μg | |
Cell Biology | |
Human | |
IgG1 |
Western Blot | |
46 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.