Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAE1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514138
Description
This antibody is predicted to react with mouse, non-human primate and rat based on sequence homology.
APPBP1 binds to the beta-amyloid precursor protein, a cell surface protein with signal-transducing properties thought to play a role in the pathogenesis of Alzheimer′s disease. In addition, this protein can form a heterodimer with UBE1C and bind and activate NEDD8, a ubiquitin-like protein. APPB1 is required for cell cycle progression through the S/M checkpoint.Specifications
NAE1 | |
Polyclonal | |
NAE1 | |
HPP1; ula-1; NAE1; A-116A10.1; NEDD8-activating enzyme E1 regulatory subunit; Amyloid protein-binding protein 1;Amyloid beta precursor protein-binding protein 1, 59 kDa; Proto-oncogene protein 1 | |
Rabbit | |
Protein G | |
RUO | |
234664, 8883 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q13564, Q8VBW6 | |
NAE1 | |
KLH conjugated synthetic peptide between 430-459 amino acids from the C-terminal region of human NAE1 (APPBP1) | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction