Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAGK Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513843

Catalog No. PIPA513843

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

N-acetylglucosamine kinase (NAGK) converts endogenous N-acetylglucosamine (GlcNAc), a major component of complex carbohydrates, from lysosomal degradation or nutritional sources into GlcNAc 6-phosphate. NAGK belongs to the group of N-acetylhexosamine kinases and is a prominent salvage enzyme of amino sugar metabolism in mammals. The predicted 344-amino acid NAGK protein contains the 5 sequence motifs necessary for the binding of ATP by sugar kinases. NAGK shares 91.6% amino acid similarity with mouse Nagk, for which enzyme activity is detectable in all mouse tissues examined, with highest enzymatic activity in testis. It is hypothesized that NAGK has a general role in the catabolic pathways of GlcNAc as well as of ManNAc.
Specifications

Specifications

NAGK
Polyclonal
PBS with 0.09% sodium azide
Q9UJ70, Q9QZ08
N-acetylglucosamine kinase; GlcNAc kinase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55577, 56174
Western Blot
Unconjugated
NAGK
Liquid
NAGK
KLH conjugated synthetic peptide between 31-61 amino acids from the N-terminal region of human NAGK
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit