Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAGLU Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528261

Catalog No. PIPA528261


Add to cart

Description

Description

PA5-28261 targets NAGLU in IF, ICC, IHC (P) and WB applications and shows reactivity with Human samples. The PA5-28261 immunogen is recombit fragment corresponding to a region within amino acids 101 and 558 of Human NAGLU. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes an enzyme that degrades heparan sulfate by hydrolysis of terminal N-acetyl-D-glucosamine residues in N-acetyl-alpha-D-glucosaminides. Defects in this gene are the cause of mucopolysaccharidosis type IIIB (MPS-IIIB), also known as Sanfilippo syndrome B. This disease is characterized by the lysosomal accumulation and urinary excretion of heparan sulfate.
Specifications

Specifications

NAGLU
Polyclonal
Unconjugated
NAGLU
Liquid
NAGLU
Recombit fragment corresponding to a region within amino acids 101 and 558 of Human NAGLU
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.69 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
P54802
MPS-IIIB, MPS3B, NAG, UFHSD, N-acetyl-alpha-glucosaminidase, alpha-N-acetylglucosaminidase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4669
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit