Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NALP6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP231372
Description
NALP6 Polyclonal specifically detects NALP6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NALP6 | |
Polyclonal | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
Angiotensin II/Vasopressin Receptor, AVR, CLR11.4, NACHT, Leucine Rich Repeat And PYD Containing 6, NACHT, LRR And PYD Containing Protein 6, NACHT, LRR And PYD Domains-Containing Protein 6, NAVR, NAVR/AVR, NLR Family, Pyrin Domain Containing 6, NLRP6 Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 6, PAN3, PYPAF5, PYRIN-Containing APAF1-Like Protein 5 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human. Immunogen sequence similarity with other species: Rat (72%), Mouse (70%) . | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1-2 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29928676), Immunohistochemistry-Paraffin 5 ug/ml | |
P59044 | |
NLRP6 | |
Partial recombinant protein made to an N-terminal portion of human NALP6 (between amino acids 10-200) [UniProt P59044]. | |
0.1 mg | |
Immunology, Inflammation | |
171389 | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction