Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAPG, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127769

Catalog No. PIPA127769

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

NSF and SNAPs are general elements of the cellular membrane transport apparatus. The sequence of the predicted 312-amino acid human protein encoded by NAPG is 95% identical to that of bovine gamma-SNAP. NAPG mediates platelet exocytosis and controls the m

PA1-27769 detects NAPG from human, mouse, and rat samples.PA1-27769 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human NAPG protein.
Specifications

Specifications

NAPG
Polyclonal
PBS with no preservative
Q99747, Q9CWZ7
GAMMASNAP
Chicken
IgY
50μg
Store at -20°C
Primary
8774
Western Blot
Unconjugated
NAPG
Purified
NAPG
Synthetic peptide corresponding to human NAPG protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit