Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NARFL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526824

Catalog No. PIPA526824

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

NARFL
Polyclonal
PBS with 0.09% sodium azide
Q9H6Q4
HPRN, IOP1, LET1L, PRN, LET1 like/JFP15, cytosolic Fe-S cluster assembly factor NARFL, iron-only hydrogenase-like protein 1, nuclear prelamin A recognition factor-like protein, protein related to Narf
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64428
Flow Cytometry, Western Blot
Unconjugated
NARFL
Liquid
NARFL
KLH conjugated synthetic peptide between 240-269 amino acids from the central region of human NARFL
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit