Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAT13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524982

Catalog No. PIPA524982


Add to cart

Description

Description

This antibody is predicted to react with bovine, mouse and Xenopus and zebrafish based on sequence homology.

NAT13 is probable catalytic component of the ARD1A-NARG1 complex which displays alpha (N-terminal) acetyltransferase activity.
Specifications

Specifications

NAT13
Polyclonal
PBS with 0.09% sodium azide
Q9GZZ1
MAK3, NAT13, NAT5, SAN, hNAT5, hSAN, Mak3 homolog, N-acetyltransferase 13 (GCN5-related), N-acetyltransferase 5, N-acetyltransferase san homolog, N-alpha-acetyltransferase 50, N-alpha-acetyltransferase 50, NatE catalytic subunit, natE catalytic subunit, separation anxiety
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80218
Flow Cytometry, Western Blot
Unconjugated
NAA50
Liquid
NAA50
KLH conjugated synthetic peptide between 117-146 amino acids from the C-terminal region of human NAT13
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit