Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAT14 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523792

Catalog No. PIPA523792


Add to cart

Description

Description

This antibody is predicted to react with bovine and non-human primate based on sequence homology.

Specifications

Specifications

NAT14
Polyclonal
Unconjugated
NAT14
Liquid
NAT14
KLH conjugated synthetic peptide between 5-33 amino acids from the N-terminal region of human NAT14
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8WUY8, Q8BVG8
KLP1, K562 cell-derived leucine-zipper-like protein 1, K562 cells-derived leucine zipper-like protein 1, K562 cells-derived leucine-zipper-like protein 1, N-acetyltransferase 14
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
269854, 57106
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit