Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAT8 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA524879
Description
This protein, isolated using the differential display method to detect tissue-specific genes, is specifically expressed in kidney and liver. The encoded protein shows amino acid sequence similarity to N-acetyltransferases. A similar protein in Xenopus affects cell adhesion and gastrulation movements, and may be localized in the secretory pathway. A highly similar paralog is found in a cluster with this gene.Specifications
NAT8 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9UHE5, Q9JIY7 | |
CML1, GLA, Hcml1, TSC501, TSC510, N-acetyltransferase 8 (camello like), camello-like protein 1, kidney- and liver-specific gene product, probable N-acetyltransferase 8 | |
Rabbit | |
IgG | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
68396, 9027 |
Immunohistochemistry, Western Blot | |
Unconjugated | |
NAT8 | |
Liquid | |
NAT8 | |
KLH conjugated synthetic peptide between 110-138 amino acids from the central region of human NAT8 | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title