Learn More
anti-Nav1.7, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP130834
Catalog No. NBP130834
Add to cart
Description
Description
Nav1.7 Polyclonal antibody specifically detects Nav1.7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Nav1.7
LYOPH
Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:300-1:2000
Q15858
ETHA, FEB3B, NE-NA, NENAVoltage-gated sodium channel subunit alpha Nav1.7, Neuroendocrine sodium channel, Peripheral sodium channel 1, PN1hNE-Na, sodium channel protein type 9 subunit alpha, Sodium channel protein type IX subunit alpha, sodium channel, voltage-gated, type IX, alpha subunit, voltage-gated sodium channel alpha subunit Nav1.7, voltage-gated, type IX, alpha polypeptide
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for SCN9A.
Human
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Whole antisera with No Preservative
Antisera
SCN9A
A synthetic peptide from the c-terminal region of human Sodium channel protein type 9 subunit alpha (SCN9A, Nav1.7) conjugated to an immunogenic carrier protein was used as the immunogen.
Unpurified
RUO
Primary
6335
Reconstitute with 0.1 ml sterilized water to desired concentration.