Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-NCALD, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP215037
Description
NCALD Polyclonal specifically detects NCALD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NCALD | |
Polyclonal | |
Unconjugated | |
0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal | |
MGC33870, MGC74858, neurocalcin delta, neurocalcin-delta | |
Rabbit | |
22 kDa | |
0.1 mL | |
Primary | |
Human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
P61601 | |
NCALD | |
Recombinant protein encompassing a sequence within the center region of human NCALD. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
83988 | |
Human, Mouse, Rat | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only