Learn More
anti-NCAM-1/CD56, PE, Clone: 809220, R&D Systems™
Manufacturer:  R&D Systems FAB7820P025
 View more versions of this product
Catalog No. FAB7820P025
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
NCAM-1/CD56
809220
Flow Cytometry 10 uL/10^6 cells
P13595
CD56, CD56/ NCAM-1, CD56 antigen, MSK39, NCAM1, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM
Rat
IgG2a
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse NCAM-1/CD56 in direct ELISAs.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
NCAM1
Mouse myeloma cell line NS0-derived recombinant mouse NCAM-1/CD56, Leu20-Thr711, Accession # P13595
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4684
Mouse