Learn More
NCAPG2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human NCAPG2 peptide using ARM Technology.
Supplier: Abnova Corporation H00054892K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Both condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 (MIM 605576) and SMC4 (MIM 605575), but they contain different sets of non-SMC subunits. NCAPG2 is 1 of 3 non-SMC subunits that define condensin II (Ono et al., 2003 [PubMed 14532007]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
NCAPG2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
NCAPG2 | |
NCAPG2 | |
A synthetic peptide of human NCAPG2 | |
RUO | |
54892 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
non-SMC condensin II complex, subunit G2 | |
NCAPG2 | |
CAP-G2, FLJ20311, LUZP5, MTB, hCAP-G2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.