Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p47phox Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525776

Catalog No. PIPA525776

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

p47phox (NCF1) is required, along with NCF2 and membrane bound cytochrome b558, for activation of the latent NADPH oxidase. This oxidase is a multicomponent enzyme that is activated to produce superoxide anion. Mutations in the gene can result in granulomatous disease.
Specifications

Specifications

p47phox
Polyclonal
PBS with 0.09% sodium azide
P14598
NCF1A, NOXO2, SH3PXD1A, p47phox, 47 kDa autosomal chronic granulomatous disease protein, 47 kDa neutrophil oxidase factor, NADPH oxidase organizer 2, NCF-1, NCF-47K, SH3 and PX domain-containing protein 1A, neutrophil NADPH oxidase factor 1, neutrophil cytosol factor 1, neutrophil cytosolic fact
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
653361
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NCF1
Liquid
NCF1
KLH conjugated synthetic peptide between 26-52 amino acids from the N-terminal region of human NCF1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit