Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCK Mouse, Unlabeled, Clone: 108, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610100

 View more versions of this product

Catalog No. BDB610100


Add to cart

Description

Description

NCK is a 47kDa protein that belongs to a class of signaling molecules called adaptors. These proteins exert their effects by coupling activated receptors to downstream effectors. NCK is comprised almost entirely of one SH2 and three SH3 domains and it possesses no intrinsic catalytic activity. Investigations of its interactions with cellular proteins have found constitutive association of NCK with various Ser/THr kinases, such as p21-associated kinase (PAK) and casein kinase 1. Other proteins that directly interact with NCK are: Sos, Cbl, Wiskott-Aldrich syndrome protein (WASP), IRS1, and multiple tyrosine kinase receptors. The presence of NCK is necessary for the mitogenic response elicited by PDGF in some cells. In addition, the Drosophila homologue of NCK is involved in regulating cytoskeletal organization.

Host Species: Mouse
Clone: 108
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Human NCK aa. 279-377
Formula Weight [Chemical]: 47kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

NCK
108
Unconjugated
Affinity Purified
Human NCK aa. 279-377
150μg
Cell Biology
Primary
Bovine, Canine, Chicken, Frog, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.