Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-NCKAP1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132339

Catalog No. NBP132339



Description

Description

NCKAP1 Polyclonal antibody specifically detects NCKAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Frozen.
Specifications

Specifications

NCKAP1
0.5mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry-Frozen 1:10 - 1:500
Q9Y2A7
FLJ11291, HEM2p125Nap1, KIAA0587, Membrane-associated protein HEM-2, MGC8981, NAP 1, NAP125, NAP1Nap1, NCK-associated protein 1
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Rat and Mouse. Predicted cross-reactivity based on sequence identity:Rhesus Monkey (100%).
Immunohistochemistry (Frozen), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
NCKAP1
Recombinant protein encompassing a sequence within the center region of human NCKAP1. The exact sequence is proprietary.
129 kDa
0.1 ml
Apoptosis
Primary
10787
Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only