Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCKAP1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525809
Description
This antibody is predicted to react with mouse, rat, Xenopus and zebrafish based on sequence homology.
Specifications
NCKAP1 | |
Polyclonal | |
NCKAP1 | |
HEM2, NAP1, NAP125, NAP 1, membrane-associated protein HEM-2, nck-associated protein 1, p125Nap1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10787 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q9Y2A7 | |
NCKAP1 | |
KLH conjugated synthetic peptide between 631-659 amino acids from the central region of human NCKAP1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction