Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-NCKIPSD, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142663

Catalog No. NBP142663


Add to cart

Description

Description

NCKIPSD Polyclonal antibody specifically detects NCKIPSD in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot (Negative).
Specifications

Specifications

NCKIPSD
1.0 mg/mL
Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Affinity Purified
NCKIPSD
The immunogen for this product maps to a region between residue 600 and 650 of human SH3 protein interacting with Nck, 90 kDa using the numbering given in entry NP_057537.1 (GeneID 51517).
Immunogen affinity purified
RUO
Primary
51517
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
54 kDa vimentin-interacting protein, 90 kDa SH3 protein interacting with Nck, AF3p21, AF3P21DIP-1, dia interacting protein, Dia-interacting protein 1, diaphanous protein interacting protein, Diaphanous protein-interacting protein, DIP, DIP1, MGC23891, NCK interacting protein with SH3 domain, ORF1, SPIN90VIP54, WASLBP, WASP-interacting SH3-domain protein, WISHSH3 adapter protein SPIN90, Wisk90 kDa
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only