Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-NCOA4, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128681

Catalog No. NBP128681


Add to cart

Description

Description

NCOA4 Polyclonal antibody specifically detects NCOA4 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

NCOA4
1.0 mg/mL
Immunohistochemistry, Immunoprecipitation 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500
NP_005428.1
Androgen receptor coactivator 70 kDa protein, Androgen receptor-associated protein of 70 kDa, ARA70RFGret fused, DKFZp762E1112, ELE1NCoA-4, nuclear receptor coactivator 4,70 kDa AR-activator, PTC3RET-activating gene ELE1, Ret-activating protein ELE1,70 kDa androgen receptor coactivator
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
NCOA4
The immunogen recognized by this antibody maps to a region between residue 500 and 550 of human 70 kDa AR-activator (nuclear receptor coactivator 4) using the numbering given in entry NP_005428.1 (GeneID 8031).
Immunogen affinity purified
RUO
Primary
8031
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only