Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCS1, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA126009

Catalog No. PIPA126009

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

This gene is a member of the neuronal calcium sensor gene family, which encode calcium-binding proteins expressed predominantly in neurons. The protein encoded by this gene regulates G protein-coupled receptor phosphorylation in a calcium-dependent manner and can substitute for calmodulin. The protein is associated with secretory granules and modulates synaptic transmission and synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.

PA1-26009 detects NCS1 from human, mouse, and rat samples.PA1-26009 has been successfully used in immunohistochemistry, and Western blot procedures.The PA1-26009 immunogen is recombinant, full length mouse protein, with extensive post-translational modification.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NCS1
Polyclonal
Unconjugated
P62166, Q8BNY6, P62168
NCS1
Recombinant, full length mouse protein, with extensive post-translational modification.
50 μg
Primary
Human, Mouse, Rat
Antibody
IgY
ELISA, Immunohistochemistry, Western Blot
0.75mg/mL
NCS1
DKFZp761L1223, FLUP, FREQ
Chicken
Antigen affinity chromatography
RUO
23413
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit