Learn More
NDRG1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523158
Catalog No. PIPA523158
Add to cart
Description
Description
NDRG1 is a 394 amino acid cytoplasmic protein belonging to the NDRG family. It interacts with APO A-I and A-II and plays a role in HDL-mediated cholesterol transport, stress responses, hormone responses, cell growth and differentiation. Its defect causes Charcot-Marie-Tooth disease type 4D (CMT4D) that is characterized by reduced nerve conduction velocities, segmental demyelination and remyelination with onion bulb formations on nerve biopsy, slowly progressive distal muscle atrophy and weakness, absent deep tendon reflexes and hollow feet. NDRG1 plays a growth inhibitory role and serves as a marker of differentiation. It is expressed in placental membranes, prostate, kidney, small intestine, ovary tissues, colon and the cells that border the lumen.
Specifications
Specifications
NDRG1
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
Q92597
CAP43, CMT4D, DRG1, GC4, HMSNL, NDR1, NMSL, PROXY1, RIT42, RTP, TARG1, TDD5, DRG-1, N-myc downstream-regulated gene 1 protein, differentiation-related gene 1 protein, nickel-specific induction protein Cap43, protein NDRG1, protein regulated by oxygen-1, reducing agents and tunicamycin-responsive protein, tunicamycin-responsive protein
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Western Blot
Unconjugated
NDRG1
Liquid
NDRG1
Synthetic peptide (382 GNSAGPKSMEVSC 394) of NDRG1 protein.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10397