Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG1 Goat anti-Human, Mouse, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA523230

Catalog No. PIPA523230


Add to cart

Description

Description

NDRG1 is a member of the emerging NDRG family involved in stress responses, hormone responses, cell growth, and differentiation. The NDRG1 gene is mapped to human chromosome 8q24.2 and encodes a cytosolic protein containing three unique tandem repeats of 10 hydrophilic amino acids near the C-terminus. Expression of NDRG1 is markedly upregulated in various cells under such conditions as cell differentiation, hypoxia and exposure to heavy metals. However, its expression is repressed by the Myc family transcription factors and after cellular transformation. Mutations in the NDRG1 gene are linked to the hereditary motor and sensory neuropathy. The protein is ubiquitously expressed in human and rat heart, brain, kidney, placenta, lung, liver, skeletal muscle, and pancreas.
Specifications

Specifications

NDRG1
Polyclonal
Unconjugated
NDRG1
Liquid
NDRG1
Synthetic peptide (382 GNSAGPKSMEVSC 394) of NDRG1 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q92597, Q62433
CAP43, CMT4D, DRG1, GC4, HMSNL, NDR1, NMSL, PROXY1, RIT42, RTP, TARG1, TDD5, DRG-1, N-myc downstream-regulated gene 1 protein, differentiation-related gene 1 protein, nickel-specific induction protein Cap43, protein NDRG1, protein regulated by oxygen-1, reducing agents and tunicamycin-responsive protein, tunicamycin-responsive protein
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10397, 17988
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit