Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527213

Catalog No. PIPA527213


Add to cart

Description

Description

PA5-27213 targets NDRG1 in Western Blot and Immunohistochemistry (Paraffin) applications and shows reactivity with Human and mouse samples. The PA5-27213 immunogen is recombit fragment corresponding to a region within amino acids 186 and 369 of Human NDRG1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein involved in stress responses, hormone responses, cell growth, and differentiation. It is necessary for p53-mediated caspase activation and apoptosis. Mutation in this gene has been reported to be causative for hereditary motor and sensory neuropathy-Lom. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications

Specifications

NDRG1
Polyclonal
Unconjugated
NDRG1
Liquid
NDRG1
Recombit fragment corresponding to a region within amino acids 186 and 369 of Human NDRG1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.67 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q92597, Q62433
CAP43, CMT4D, DRG1, GC4, HMSNL, NDR1, NMSL, PROXY1, RIT42, RTP, TARG1, TDD5, DRG-1, N-myc downstream-regulated gene 1 protein, differentiation-related gene 1 protein, nickel-specific induction protein Cap43, protein NDRG1, protein regulated by oxygen-1, reducing agents and tunicamycin-responsive protein, tunicamycin-responsive protein
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10397, 17988
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit