Learn More
NDRG2, Mouse, Clone: 1D6, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant NDRG2.
Supplier: Abnova Corporation H00057447M10
Description
This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq
Sequence: MAELQEVQITEEKPLLPGQTPEAAKEAELAARILLDQGQTHSVETPYVSVTFTVYGTPKPKRPAILTYHDVGLNYKSCFQPLFQFEDMQEIIQNFVRVHVDAPGMEEGAPVFPLGYQYPSLDQLADMIPCVLQYLNFSTIIGVGVGAGAYILARYALNHPDTVEGLVLINIDPNAKGWMDWAAHKLTGLTSSIPEMILGHLFSQEELSGNSELIQKYRNIITHAPNLDNIELYWNSYNNRRDLNFERGGDITLRCPVMLVVECNSKLDPTQTSFLKMADSGGQPQLTQPGKLTEAFKYFLQGMGYMASSCMTRLSRSRTASLTSAASVDGNRSRSRTLSQSSESGTLSSGPPGHTMEVSCSpecifications
NDRG2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC011240 | |
NDRG2 | |
NDRG2 (AAH11240, 1 a.a. ∼ 360 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
1D6 | |
Mouse monoclonal antibody raised against a full-length recombinant NDRG2. | |
NDRG2 | |
DKFZp781G1938/FLJ25522/KIAA1248/SYLD | |
Mouse | |
Affinity chromatography | |
RUO | |
57447 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.