Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDST2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525662

Catalog No. PIPA525662

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

NDST2 is a member of theN-deacetylase/N-sulfotransferase subfamily of the sulfotransferase1 proteins. The encoded enzyme has dual functions in processing glucosamine and heparin polymers, including N-deacetylation andN-sulfation. The encoded protein may be localized to the Golgi.
Specifications

Specifications

NDST2
Polyclonal
PBS with 0.09% sodium azide
P52849, P52850
HSST2, NST2, N-HSST 2, N-heparan sulfate sulfotransferase 2, NDST-2, bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 2, glucosaminyl N-deacetylase/N-sulfotransferase 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
17423, 8509
Western Blot
Unconjugated
NDST2
Liquid
NDST2
KLH conjugated synthetic peptide between 817-844 amino acids from the C-terminal region of human NDST2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit