Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA11 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524022

Catalog No. PIPA524022


Add to cart

Description

Description

NDUFA11 encodes a subunit of the membrane-bound mitochondrial complex I. Complex I is composed of numerous subunits and functions as the NADH-ubiquinol reductase of the mitochondrial electron transport chain. Mutations in this gene are associated with severe mitochondrial complex I deficiency. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

NDUFA11
Polyclonal
Unconjugated
NDUFA11
Liquid
NDUFA11
KLH conjugated synthetic peptide between 64-92 amino acids from the central region of human NDUAB
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q86Y39
B14.7, CI-B14.7, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 11, NADH-ubiquinone oxidoreductase subunit B14.7, complex I B14.7 subunit
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
126328
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit