Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFAF3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524469

Catalog No. PIPA524469

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a mitochondrial complex I assembly protein that interacts with complex I subunits. Mutations in this gene cause mitochondrial complex I deficiency, a fatal neonatal disorder of the oxidative phosphorylation system. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications

Specifications

NDUFAF3
Polyclonal
PBS with 0.09% sodium azide
Q9BU61, Q9JKL4
2P1, C3orf60, E3-3, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 3, nuclear protein E3-3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25915, 66706
Immunohistochemistry, Western Blot
Unconjugated
NDUFAF3
Liquid
NDUFAF3
KLH conjugated synthetic peptide between 12-40 amino acids from the N-terminal region of human NDUF3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit