Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA10 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526771

Catalog No. PIPA526771

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene belongs to the complex I 42kDA subunit family. Mammalian complex I is the first enzyme complex in the electron transport chain of mitochondria. It is composed of 45 different subunits. This protein is a component of the hydrophobic protein fraction and has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain.
Specifications

Specifications

NDUFA10
Polyclonal
PBS with 0.09% sodium azide
O95299, Q99LC3
CI-42KD, CI-42k, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 10, mitochondrial, NADH-ubiquinone oxidoreductase 42 kDa subunit, complex I 42kDa subunit
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4705, 67273
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
NDUFA10
Liquid
NDUFA10
KLH conjugated synthetic peptide between 252-281 amino acids from the central region of human NDUFA10
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit