Learn More
anti-NDUFA4, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232318
Catalog No. NBP232318
Add to cart
Description
Description
NDUFA4 Polyclonal antibody specifically detects NDUFA4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
NDUFA4
0.5 mg/mL
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500
O00483
CI-9k, CI-MLRQ, Complex I 9kDa subunit, Complex I-MLRQ, FLJ27440, MGC104422, MGC126843, MGC126845, MLRQ, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4 (9kD, MLRQ), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4, 9kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4, NADH-ubiquinone oxidoreductase MLRQ subunit
Rabbit
IgG
Protein A purified
RUO
Primary
4697
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
NDUFA4
This NDUA4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 56-80 amino acids from the C-terminal region of human NDUA4.
9370 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human