Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFAB1, Mouse, MaxPab™ Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human NDUFAB1 protein.

Manufacturer:  Abnova Corporation H00004706B01

Catalog No. 89-002-693


Add to cart

Description

Description

Sequence: MASRVLSAYVSRLPAAFAPLPRVRMLAVARPLSTALCSAGTQTRLGTLQPALVLAQVPGRVTQLCRQYSDMPPLTLEGIQDRVLYVLKLYDKIDPEKLSVNSHFMKDLGLDSLDQVEIIMAMEDEFGFEIPDIDAEKLMCPQEIVDYIADKKDVYE
Specifications

Specifications

NDUFAB1
Unconjugated
No additive
NM_005003.2
ACP|FASN2A|MGC65095|SDAP
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human NDUFAB1 protein.
NDUFAB1
Liquid
NDUFAB1
NDUFAB1 (NP_004994.1, 1 a.a. ~ 156 a.a) full-length human protein.
RUO
Primary
4706
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit