Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFAF1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524236

Catalog No. PIPA524236

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The NDUFAF1 gene encodes the human homolog of a Neurospora crassa Cia30 protein involved in the assembly of complex I.
Specifications

Specifications

NDUFAF1
Polyclonal
PBS with 0.09% sodium azide
Q9Y375
CGI65, CIA30, NADH-ubiquinone oxidoreductase 1 alpha subcomplex, assembly factor 1, complex I intermediate-associated protein 30, mitochondrial
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51103
Immunohistochemistry, Western Blot
Unconjugated
NDUFAF1
Liquid
NDUFAF1
KLH conjugated synthetic peptide between 247-276 amino acids from the C-terminal region of human NDUFAF1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit