Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFB4 Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA529208

Catalog No. PIPA529208


Add to cart

Description

Description

PA5-29208 targets NDUFB4 in IHC (P) and WB applications and shows reactivity with Human and Rat samples. The PA5-29208 immunogen is recombit fragment corresponding to a region within amino acids 1 and 129 of Human NDUFB4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene belongs to the complex I NDUFB4 subunit family. Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications

Specifications

NDUFB4
Polyclonal
Unconjugated
NDUFB4
Liquid
NDUFB4
Recombit fragment corresponding to a region within amino acids 1 and 129 of Human NDUFB4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
O95168
B15, CI-B15, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 4, NADH-ubiquinone oxidoreductase B15 subunit, complex I B15 subunit, complex I-B15
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
288088, 4710
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit