Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFC2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524234

Catalog No. PIPA524234

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications

Specifications

NDUFC2
Polyclonal
PBS with 0.09% sodium azide
O95298
B14.5b, CI-B14.5b, HLC-1, NADHDH2, NADH dehydrogenase [ubiquinone] 1 subunit C2, NADH-ubiquinone oxidoreductase subunit B14.5b, complex I subunit B14.5b, complex I-B14.5b, human lung cancer oncogene 1 protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4718
Immunohistochemistry, Western Blot
Unconjugated
NDUFC2
Liquid
NDUFC2
KLH conjugated synthetic peptide between 90-119 amino acids from the C-terminal region of human NDUFC2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit