Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFS2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522364

Catalog No. PIPA522364


Add to cart

Description

Description

PA5-22364 targets NDUFS2 in IHC (P) and WB applications and shows reactivity with Human and mouse samples. The PA5-22364 immunogen is a recombit fragment corresponding to a region within amino acids 200 and 430 of NDUFS2 (Uniprot IDNo.O75306). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

NDUFS2
Polyclonal
Unconjugated
NDUFS2
Liquid
NDUFS2
Recombit fragment corresponding to a region within amino acids 200 and 430 of NDUFS2 (Uniprot IDNo.O75306)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.89 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
O75306, Q91WD5
CI-49, CI-49kD, NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial, NADH-ubiquinone oxidoreductase 49 kDa subunit, NADH-ubiquinone oxidoreductase NDUFS2 subunit, complex 1, mitochondrial respiratory chain, 49-KD subunit, complex I 49kDa subunit, complex I-49kD
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
226646, 4720
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit