Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFS3, Mouse, MaxPab™ Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human NDUFS3 protein.

Manufacturer:  Abnova Corporation H00004722B01

Catalog No. 89-002-694


Add to cart

Description

Description

This gene encodes one of the iron-sulfur protein (IP) components of mitochondrial NADH:ubiquinone oxidoreductase (complex I). Mutations in this gene are associated with Leigh syndrome resulting from mitochondrial complex I deficiency

Sequence: MAAAAVARLWWRGILGASALTRGTGRPSVLLLPVRRESAGADTRPTVRPRNDVAHKQLSAFGEYVAEILPKYVQQVQVSCFNELEVCIHPDGVIPVLTFLRDHTNAQFKSLVDLTAVDVPTRQNRFEIVYNLLSLRFNSRIRVKTYTDELTPIESAVSVFKAANWYEREIWDMFGVFFANHPDLRRILTDYGFEGHPFRKDFPLSGYVELRYDDEVKRVVAEPVELAQEFRKFDLNSPWEAFPVYRQPPESLKLEAGDKKPDAK
Specifications

Specifications

NDUFS3
Unconjugated
No additive
NM_004551.1
NDUFS3
NDUFS3 (NP_004542.1, 1 a.a. ~ 264 a.a) full-length human protein.
RUO
Primary
4722
Western Blot
Mouse polyclonal antibody raised against a full-length human NDUFS3 protein.
NDUFS3
Liquid
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit