Learn More
NDUFS4 Mouse anti-Human, Clone: 3F11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519432
Catalog No. PIMA519432
Add to cart
Description
Description
Peptide Sequence: GVPEEHIKTR KVRIFVPARN NMQSGVNNTK KWKMEFDTRE RWENPLMGWA STADPLSNMV LTFSTKEDAV SFAEKNGWSY DIEERKVPKP KSKSYGANFS WNKRTRVSTK
This gene encodes an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), or NADH:ubiquinone oxidoreductase, the first multi-subunit enzyme complex of the mitochondrial respiratory chain. Complex I plays a vital role in cellular ATP production, the primary source of energy for many crucial processes in living cells. It removes electrons from NADH and passes them by a series of different protein-coupled redox centers to the electron acceptor ubiquinone. In well-coupled mitochondria, the electron flux leads to ATP generation via the building of a proton gradient across the inner membrane. Complex I is composed of at least 41 subunits, of which 7 are encoded by the mitochondrial genome and the remainder by nuclear genes.
Specifications
Specifications
NDUFS4
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
O43181
AQDQ, CI-18, CI-18 kDa, CI-AQDQ, NADH dehydrogenase (ubiquinone) iron-sulfur protein 4, NADH dehydrogenase [ubiquinone] iron-sulfur protein 4, mitochondrial, NADH-coenzyme Q reductase, 18-KD, NADH-ubiquinone oxidoreductase 18 kDa subunit, complex I 18kDa subunit, complex I-18 kDa, complex I-AQDQ, mitochondrial respiratory chain complex I (18-KD subunit)
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4724
ELISA, Western Blot
3F11
Unconjugated
NDUFS4
Liquid
NDUFS4
NDUFS4 (NP_002486, 66 a.a. ∽ 175 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human