Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDV (Newcastle disease virus ), Mouse, Clone: 9F7, Abnova™

Mouse monoclonal antibody raised against Newcastle disease virus, La-sota strain.

Manufacturer:  Abnova Corporation MAB0572

Catalog No. 89-024-819


Add to cart

Specifications

Specifications

NDV (Newcastle disease virus )
9F7
Mouse monoclonal antibody raised against Newcastle disease virus, La-sota strain.
In PBS, pH 7.4 (0.09% sodium azide)
Murine
IgG1
RUO
Primary
ELISA, Inhibition Assays
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
La-sota strain.
200 ug
Store at 4°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit