Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEDD4 Rabbit anti-Human, Equine, Monkey, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178067

Catalog No. NBP178067

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

NEDD4 Polyclonal antibody specifically detects NEDD4 in Human, Equine, Monkey samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

NEDD4
0.64 mg/ml
Western Blot 1:300-1:500, ELISA 1:20000 – 1:35000, Immunocytochemistry/Immunofluorescence 1:10-1:500
NP_006145
Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1
Rabbit
IgG
0.1 mg
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
This antibody reacts with endogenous Nedd4 protein. A BLAST analysis was used to suggest reactivity with Nedd4 from human, horse and macaque based on a 100% homology.
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
NEDD4
This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a peptide corresponding to an internal portion of the NEDD1 protein.
Immunogen affinity purified
RUO
Primary
4734
Equine, Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only