Learn More
MilliporeSigma™ anti-Nedd4, WW2 domain; Polyclonal; ™ (Upstate™)
Supplier: MilliporeSigma™ 07049/DEL
Description
Staub, O., et al., American Journal of Physiology, 272: C1871-80, 1997.
Immunohistochemistry, Western Blotting
Specifications
Nedd4, WW2 domain | |
Unconjugated | |
GST fusion protein corresponding to the WW2 domain (residues 395-462) of rat Nedd4 | |
RUO | |
Primary | |
Semi-purified |
Polyclonal | |
Rabbit | |
200 μL | |
Protein Trafficking | |
Human, Mouse, Rat | |