Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

NEDD9, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant NEDD9.

Manufacturer:  Abnova Corporation H00004739A01

Catalog No. 89-003-040



Description

Description

Sequence: PRDTIYQVPPSYQNQGIYQVPTGHGTQEQEVYQVPPSVQRSIGGTSGPHVGKKVITPVRTGHGYVYEYPSRYQKDVYDIPPSHTTQGVYDIPPSSAKGPV
Specifications

Specifications

NEDD9
Unconjugated
50% glycerol
NM_006403
CAS-L|CAS2|CASL|CASS2|HEF1|dJ49G10.2|dJ761I2.1
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant NEDD9.
NEDD9
Liquid
NEDD9
NEDD9 (NP_006394, 101 a.a. ~ 200 a.a) partial recombinant protein with GST tag.
RUO
Primary
4739
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit