Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEIL3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14304

Catalog No. PA14304


Add to cart

Description

Description

PA1-4304 detects the human NEI3 protein in human samples. PA1-4304 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~68 kDa protein representing NEI3 in human testis lysate. PA1-4304 immunizing peptide corresponds to amino acid residues 164-177 of the human NEI3 protein. There is 85% sequence identity to mouse and rat.

Prokaryotes and lower eukaryotes possess redundant activities that remove the plethora of oxidative DNA base damages produced during normal oxidative metabolism and which have been associated with cancer and aging. Thus far, only one oxidized pyrimidine-specific DNA glycosylase has been identified in humans, hNthl. HNEI3 is one of three new putative human DNA glycosylases.
Specifications

Specifications

NEIL3
Polyclonal
Unconjugated
NEIL3
Liquid
NEIL3
Synthetic peptide: L(164) R A E S E V K K Q K G R M L G(177)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8TAT5
FGP2, FLJ10858, FPG2, NEI3, hFPG2, hNEI3
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
55247
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit