Learn More
Neisseria gonorrhoeae Rabbit anti-Bacteria, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA173145
Catalog No. PIPA173145
Add to cart
Description
Description
The PA1-73145 antibody reacts with Neisseria gonorrhoeae. PA1-73145 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73145 antibody was raised against whole N. gonorrhoeae; ATCC 31426.
The genus Neisseria contains a number of species which are normal flora and pathogens of humans and animals. Of these species, the species of human origin, and particularly the pathogenic species, N. gonorrhoeae and N. meningitidis, have been studied extensively in an effort to control the infections they cause. Gonorrhea, caused by N. gonorrhoeae, is one of the most frequently reported infectious diseases in the United States and worldwide. Of the Neisseria and related species, only N. gonorrhoeae is considered always to be pathogenic i.e., cause disease. N. gonorrhoeae is not considered to be normal flora under any circumstances.
Specifications
Specifications
Neisseria gonorrhoeae
Polyclonal
Biotin
Liquid
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Bacteria
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
4-5 mg/mL
PBS with 0.1% sodium azide
N. gonorrhoeae; Gonococci
Whole N. gonorrhoeae; ATCC 31426.
Purified
RUO
Antibody
Polyclonal