Learn More
Neisseria gonorrhoeae, FITC, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA127302
Catalog No. PA127302
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The genus Neisseria contains a number of species which are normal flora and pathogens of humans and animals. Of these species, the species of human origin, and particularly the pathogenic species, N. gonorrhoeae and N. meningitidis, have been studied extensively in an effort to control the infections they cause. Gonorrhea, caused by N. gonorrhoeae, is one of the most frequently reported infectious diseases in the United States and worldwide. Of the Neisseria and related species, only N. gonorrhoeae is considered always to be pathogenic i.e., cause disease. N. gonorrhoeae is not considered to be normal flora under any circumstances.
PA1-27302 detects Neisseria gonorrhea from bacteria samples.PA1-27302 has been successfully used in immunofluorescence and ELISA procedures.The PA1-27302 immunogen is whole N. gonorrhoeae; ATCC 31426.Store at 4ºC short term. For extended storage, aliquot and store at -20ºC or below. Avoid freeze/thaw cycles.
Specifications
Specifications
Neisseria gonorrhoeae
Polyclonal
FITC
Purified
Rabbit
IgG
100μL
Store at -20°C
Primary
Bacteria
ELISA, Immunocytochemistry, Immunofluorescence
4mg/mL
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
N. gonorrhoeae, Gonococci
Whole N. gonorrhoeae; ATCC 31426.
IgG fraction
RUO
Antibody
Polyclonal