Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK11 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515334

Catalog No. PIPA515334

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

NEK11 belongs to the NIMA family of kinases, which are involved in DNA replication and genotoxic stress responses.
Specifications

Specifications

NEK11
Polyclonal
PBS with 0.09% sodium azide
Q8NG66, Q8C0Q4
NIMA-related kinase 11L
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
208583, 79858
Western Blot
Unconjugated
NEK11
Liquid
NEK11
KLH conjugated synthetic peptide between 480-510 amino acids from the C-terminal region of human NEK11L
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit