Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515341

Catalog No. PIPA515341

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rat based on sequence homology.

NIMA-related kinases share high amino acid sequence identity with the gene product of the Aspergillus nidulans  ft.never in mitosis A ft. gene, which controls initiation of mitosis.[supplied by OMIM]
Specifications

Specifications

NEK7
Polyclonal
PBS with 0.09% sodium azide
Q8TDX7
Serine/threonine-protein kinase Nek7, NIMA (never in mitosis gene a)-related kinase 7, NimA-related protein kinase 7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
140609
Immunohistochemistry, Western Blot
Unconjugated
NEK7
Liquid
NEK7
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human NEK7
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit