Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513864

Catalog No. PIPA513864

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

NEK8 is a member of the serine/threionine protein kinase family related to NIMA (never in mitosis, gene A) of Aspergillus nidulans. The encoded protein may play a role in cell cycle progression from G2 to M phase. Mutations in the related mouse gene are associated with a disease phenotype that closely parallels the juvenile autosomal recessive form of polycystic kidney disease in humans.
Specifications

Specifications

NEK8
Polyclonal
PBS with 0.09% sodium azide
Q86SG6
Serine/threonine-protein kinase Nek8; NimA-related protein kinase 8; NIMA-related kinase 12a; NEK12A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
284086
Western Blot
Unconjugated
NEK8
Liquid
NEK8
KLH conjugated synthetic peptide between 65-95 amino acids from the N-terminal region of human NEK8
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit