Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NELL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527958

Catalog No. PIPA527958


Add to cart

Description

Description

PA5-27958 targets NELL1 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27958 immunogen is recombit fragment corresponding to a region within amino acids 50 and 505 of Human NELL1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF)-like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. Two transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

NELL1
Polyclonal
Unconjugated
NELL1
Liquid
NELL1
Recombit fragment corresponding to a region within amino acids 50 and 505 of Human NELL1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.85 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q92832
IDH3GL, NRP1, NEL-like protein 1, nel-related protein 1, neural epidermal growth factor-like 1, protein kinase C-binding protein NELL1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4745
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit