Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-NELL2, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals 46030002
Description
NELL2 Polyclonal antibody specifically detects NELL2 in Human samples. It is validated for Western Blot, ELISA.Specifications
NELL2 | |
Unconjugated | |
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative | |
Affinity Purified | |
NELL2 | |
This antibody was made against a protein fragment from the Middle Region | |
Immunogen affinity purified | |
RUO | |
Primary | |
4753 | |
Human |
ELISA, Western Blot | |
Western Blot 1:5000-1:20000, ELISA 1:100-1:2000 | |
Q99435 | |
nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2 | |
Rabbit | |
IgG | |
0.05 mg | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
This product is specific for Human NELL2. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only